ENA413166810 Enamine

2-(hydroxymethyl)pyridine-3-carboxamide

  • MDL number MFCD18802494
  • Cas Number : 115012-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142586
Availability details for
Quantity
  • Availability
  • Add to Cart