ENA413166812 Enamine

6-butoxypyridin-3-amine

  • MDL number MFCD00023469
  • Cas Number : 539-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142589
Availability details for
Quantity
  • Availability
  • Add to Cart