ENA413166817 Enamine

3-chloro-N-ethylpyrazin-2-amine

  • MDL number MFCD16079942
  • Cas Number : 53265-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142610

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142610
Availability details for
Quantity
  • Availability
  • Add to Cart