ENA413166984 Enamine

5-[2-(trifluoromethyl)phenyl]-1H-pyrazol-3-amine

  • MDL number MFCD20486776
  • Cas Number : 502133-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-145049
Availability details for
Quantity
  • Availability
  • Add to Cart