ENA413166992 Enamine

ethyl 2-phenylpent-4-enoate

  • MDL number MFCD27500732
  • Cas Number : 14815-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142622
Availability details for
Quantity
  • Availability
  • Add to Cart