ENA413167036 Enamine

tert-butyl 4-(methylamino)butanoate

  • MDL number MFCD08275842
  • Cas Number : 138007-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145268

Safety Information

msds https://www.enaminestore.com/catalog/EN300-145268
Availability details for
Quantity
  • Availability
  • Add to Cart