ENA413167123 Enamine

2-methyl-4-[(2R)-pyrrolidin-2-yl]-1,3-thiazole

  • MDL number MFCD18643454
  • Cas Number : 1272728-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142628
Availability details for
Quantity
  • Availability
  • Add to Cart