ENA413167131 Enamine

(3-methylbutan-2-yl)({[1,2,4]triazolo[4,3-a]pyridin-3-yl}methyl)amine

  • MDL number MFCD11162709
  • Cas Number : 1042524-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-145428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-145428
Availability details for
Quantity
  • Availability
  • Add to Cart