ENA413167266 Enamine

piperidin-3-ylmethanesulfonamide

  • MDL number MFCD14585293
  • Cas Number : 1206969-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145642

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-145642
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart