ENA413167410 Enamine

1-(1H-1,3-benzodiazol-2-yl)piperidin-4-amine

  • MDL number MFCD12768755
  • Cas Number : 933698-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-145849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-145849
Availability details for
Quantity
  • Availability
  • Add to Cart