ENA413167435 Enamine

1-[1-(3-fluorophenyl)ethyl]piperazine

  • MDL number MFCD05187168
  • Cas Number : 516447-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-145891
Availability details for
Quantity
  • Availability
  • Add to Cart