ENA413167437 Enamine

methyl 2-amino-2-methyl-3-phenoxypropanoate

  • MDL number MFCD16143032
  • Cas Number : 1250200-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-145893
Availability details for
Quantity
  • Availability
  • Add to Cart