ENA413167454 Enamine

3-ethoxy-4-methylaniline

  • MDL number MFCD11934371
  • Cas Number : 2486-64-8
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-145917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-145917
Availability details for
Quantity
  • Availability
  • Add to Cart