ENA413167490 Enamine

4-ethyl-1,3-thiazol-2-amine

  • MDL number MFCD13179620
  • Cas Number : 34631-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145993

Safety Information

msds https://www.enaminestore.com/catalog/EN300-145993
Availability details for
Quantity
  • Availability
  • Add to Cart