ENA413167494 Enamine

8-(aminomethyl)-1,4-dioxaspiro[4.5]decan-8-ol

  • MDL number MFCD12169412
  • Cas Number : 863565-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-145998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-145998
Availability details for
Quantity
  • Availability
  • Add to Cart