ENA413167788 Enamine

4-(cyclopropylmethoxy)-2-fluoroaniline

  • MDL number MFCD16074843
  • Cas Number : 1236764-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-146485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-146485
Availability details for
Quantity
  • Availability
  • Add to Cart