ENA413167804 Enamine

1-bromonaphthalen-2-amine

  • MDL number MFCD00086888
  • Cas Number : 20191-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-146502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-146502
Availability details for
Quantity
  • Availability
  • Add to Cart