ENA413168180 Enamine

[2-(difluoromethoxy)-6-fluorophenyl]methanamine

  • MDL number MFCD12088134
  • Cas Number : 1153055-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-147283
Availability details for
Quantity
  • Availability
  • Add to Cart