ENA413168185 Enamine

3-(tert-butoxy)azetidine

  • MDL number MFCD16871882
  • Cas Number : 1147530-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147298
Availability details for
Quantity
  • Availability
  • Add to Cart