ENA413168297 Enamine

methyl 2-(cyclopropylamino)acetate

  • MDL number MFCD11149654
  • Cas Number : 62813-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147621
Availability details for
Quantity
  • Availability
  • Add to Cart