ENA413168298 Enamine

N-methylpiperazine-1-carboxamide

  • MDL number MFCD16300787
  • Cas Number : 163361-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147628
Availability details for
Quantity
  • Availability
  • Add to Cart