ENA413168399 Enamine

6-methyl-2-(trifluoromethyl)pyrimidine-4-carboxylic acid

  • MDL number MFCD19440056
  • Cas Number : 945717-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147906
Availability details for
Quantity
  • Availability
  • Add to Cart