ENA413168417 Enamine

N-(piperidin-4-yl)pyridin-3-amine

  • MDL number MFCD12778508
  • Cas Number : 63260-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-147973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-147973
Availability details for
Quantity
  • Availability
  • Add to Cart