ENA413168697 Enamine

methyl[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)methyl]amine

  • MDL number MFCD16860176
  • Cas Number : 1249712-29-5
  • Molecular Weight : 127.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl(1-methyl-1H-tetraazol-5-yl)methanamine
Inchi Code 1S/C4H9N5/c1-5-3-4-6-7-8-9(4)2/h5H,3H2,1-2H3
Inchi Key UCMFGLAEQNQETA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-148515

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-148515
pictograms GHS05,GHS07
hstatements H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart