ENA413168735 Enamine

5,7-dichloro-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD08544287
  • Cas Number : 89315-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-148593

Safety Information

msds https://www.enaminestore.com/catalog/EN300-148593
Availability details for
Quantity
  • Availability
  • Add to Cart