ENA413168753 Enamine

4-(4-butoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD00440590
  • Cas Number : 15850-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-148650

Safety Information

msds https://www.enaminestore.com/catalog/EN300-148650
Availability details for
Quantity
  • Availability
  • Add to Cart