ENA413168889 Enamine

2-[2-(methylamino)ethyl]-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD14656030
  • Cas Number : 1249944-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-148948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-148948
Availability details for
Quantity
  • Availability
  • Add to Cart