ENA413168971 Enamine

2-({1-[(tert-butoxy)carbonyl]piperidin-4-yl}sulfanyl)acetic acid

  • MDL number MFCD27500733
  • Cas Number : 766539-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-150771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-150771
Availability details for
Quantity
  • Availability
  • Add to Cart