ENA413169221 Enamine

[3-(methylamino)phenyl]methanol

  • MDL number MFCD16658585
  • Cas Number : 121562-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-149541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-149541
Availability details for
Quantity
  • Availability
  • Add to Cart