ENA413169342 Enamine

3,3-dimethyl-4-(methylamino)butan-1-ol

  • MDL number MFCD19374422
  • Cas Number : 1268153-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-149999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-149999
Availability details for
Quantity
  • Availability
  • Add to Cart