ENA413169350 Enamine

N-[2-(methylamino)ethyl]acetamide

  • MDL number MFCD10001503
  • Cas Number : 4814-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-150022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-150022
Availability details for
Quantity
  • Availability
  • Add to Cart