ENA413169528 Enamine

3-methylimidazolidin-4-one

  • MDL number MFCD19223268
  • Cas Number : 131213-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-150358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-150358
Availability details for
Quantity
  • Availability
  • Add to Cart