ENA413169561 Enamine

5-methyl-2-(piperidin-4-yloxy)pyrimidine

  • MDL number MFCD18293029
  • Cas Number : 1211584-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-150411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-150411
Availability details for
Quantity
  • Availability
  • Add to Cart