ENA413169617 Enamine

2-[(1H-1,2,3-triazol-1-yl)methyl]piperidine

  • MDL number MFCD27920171
  • Cas Number : 1432792-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-150507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-150507
Availability details for
Quantity
  • Availability
  • Add to Cart