ENA413169713 Enamine

3-(2H-1,2,3-triazol-4-yl)aniline

  • MDL number MFCD09035300
  • Cas Number : 876343-39-4
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2H-1,2,3-triazol-4-yl)aniline
Inchi Code 1S/C8H8N4/c9-7-3-1-2-6(4-7)8-5-10-12-11-8/h1-5H,9H2,(H,10,11,12)
Inchi Key WSRYWQPIUCQKLN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-150728

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-150728
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart