ENA413169741 Enamine

5-methyl-1-[(oxan-4-yl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD20330848
  • Cas Number : 1340093-64-2
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1-(tetrahydro-2H-pyran-4-ylmethyl)-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C11H16N2O3/c1-8-10(11(14)15)6-12-13(8)7-9-2-4-16-5-3-9/h6,9H,2-5,7H2,1H3,(H,14,15)
Inchi Key JWRRBCDGUBJUFY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-150810

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-150810
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart