ENA413169912 Enamine

dimethyl[(4-methylpiperidin-4-yl)methyl]amine

  • MDL number MFCD18271760
  • Cas Number : 1343094-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-151479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-151479
Availability details for
Quantity
  • Availability
  • Add to Cart