ENA413169922 Enamine

6-(propan-2-yl)-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD20318956
  • Cas Number : 935655-08-6
  • Molecular Weight : 175.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-isopropyl-1,2,3,4-tetrahydroisoquinoline
Inchi Code 1S/C12H17N/c1-9(2)10-3-4-12-8-13-6-5-11(12)7-10/h3-4,7,9,13H,5-6,8H2,1-2H3
Inchi Key SSWUUFFQRQRUNJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-151508

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P312,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-151508
pictograms GHS07
hstatements H303,H315,H319,H333,H335
Availability details for
Quantity
  • Availability
  • Add to Cart