ENA413170139 Enamine

2-[1-(methoxymethyl)cyclopropyl]ethanimidamide hydrochloride

  • MDL number MFCD27500738
  • Cas Number : 1803608-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152300
Availability details for
Quantity
  • Availability
  • Add to Cart