ENA413170141 Enamine

methyl 2-(5-methyl-1,3,4-oxadiazol-2-yl)acetate

  • MDL number MFCD27920197
  • Cas Number : 19813-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152321

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-152321
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart