ENA413170147 Enamine

potassium 5-(2-methylcyclopropyl)-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD27500742
  • Cas Number : 1803580-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-152414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152414
Availability details for
Quantity
  • Availability
  • Add to Cart