ENA413170152 Enamine

methyl 3-amino-3-methylbutanoate hydrochloride

  • MDL number MFCD27920198
  • Cas Number : 69950-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152514
Availability details for
Quantity
  • Availability
  • Add to Cart