ENA413170157 Enamine

N-[2-(3-aminobenzenesulfonamido)ethyl]acetamide hydrochloride

  • MDL number MFCD27500757
  • Cas Number : 1803599-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152602
Availability details for
Quantity
  • Availability
  • Add to Cart