ENA413170174 Enamine

2,2,2-trifluoroethyl 2-chloro-2-oxoacetate

  • MDL number MFCD26936297
  • Cas Number : 958649-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152754

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152754
Availability details for
Quantity
  • Availability
  • Add to Cart