ENA413170182 Enamine

methyl 5-methyl-3H-imidazo[4,5-b]pyridine-7-carboxylate; trifluoroacetic acid

  • MDL number MFCD27500785
  • Cas Number : 1803589-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152844
Availability details for
Quantity
  • Availability
  • Add to Cart