ENA413170185 Enamine

3-(4-aminobutyl)-1,3-diazaspiro[4.4]nonane-2,4-dione hydrochloride

  • MDL number MFCD27500790
  • Cas Number : 1803606-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152906
Availability details for
Quantity
  • Availability
  • Add to Cart