ENA413170187 Enamine

(2-aminoethyl)(methoxy)methylamine dihydrochloride

  • MDL number MFCD27500792
  • Cas Number : 1803599-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-152909
Availability details for
Quantity
  • Availability
  • Add to Cart