ENA413170192 Enamine

methyl 2-(3,4-dichlorophenyl)-2-(methylamino)propanoate hydrochloride

  • MDL number MFCD27500798
  • Cas Number : 1803582-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153018
Availability details for
Quantity
  • Availability
  • Add to Cart