ENA413170200 Enamine

4-(4-hydroxyphenoxy)-3-methylbenzoic acid

  • MDL number MFCD27500807
  • Cas Number : 93044-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153057
Availability details for
Quantity
  • Availability
  • Add to Cart