ENA413170201 Enamine

[amino(3-fluorophenyl)methylidene]amino 2-chloropropanoate

  • MDL number MFCD27500808
  • Cas Number : 1807941-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-153062
Availability details for
Quantity
  • Availability
  • Add to Cart